ENA947393142 Enamine

methyl 2-imino-1,2-dihydroquinazoline-5-carboxylate

  • MDL number MFCD31583506
  • Cas Number : 2126161-00-8
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-373307

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-373307
Availability details for
Quantity
  • Availability
  • Add to Cart