ENA947393192 Enamine

5-(bromomethyl)-4-iodo-1-methyl-1H-pyrazole hydrobromide

  • MDL number MFCD30749968
  • Cas Number : 2126178-39-8
  • Molecular Weight : 381.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-345992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-345992
Availability details for
Quantity
  • Availability
  • Add to Cart