ENA947393198 Enamine

methyl N-(4-cyano-3-methyl-1,2-thiazol-5-yl)-N-propylcarbamate

  • MDL number MFCD31381312
  • Cas Number : 2126162-39-6
  • Molecular Weight : 239.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-365301

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-365301
Availability details for
Quantity
  • Availability
  • Add to Cart