ENA947397431 Enamine

9-benzyl-7-methyl-6-oxa-2,9-diazaspiro[4.5]decane

  • MDL number MFCD31381322
  • Cas Number : 2126159-81-5
  • Molecular Weight : 246.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-378367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378367
Availability details for
Quantity
  • Availability
  • Add to Cart