ENA947397581 Enamine

2-chloro-2-fluoroacetyl chloride

  • MDL number MFCD01861134
  • Cas Number : 359-32-0
  • Molecular Weight : 130.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-378606

Safety Information

signalword Danger
MSDS https://www.enaminestore.com/catalog/EN300-378606
pictograms GHS02,GHS05,GHS07
hstatements EUH014,H225,H290,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart