ENA947400518 Enamine

1-cyclopropyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD19313330
  • Cas Number : 1267956-49-9
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-637412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-637412
Availability details for
Quantity
  • Availability
  • Add to Cart