ENA947402008 Enamine

3-cyclohexylpropane-1-sulfonyl chloride

  • MDL number MFCD19201134
  • Cas Number : 195053-79-3
  • Molecular Weight : 224.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-681246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-681246
Availability details for
Quantity
  • Availability
  • Add to Cart