ENA947402261 Enamine

1-{[(9H-fluoren-9-yl)methoxy]carbonyl}-2,3-dihydro-1H-indole-3-carboxylic acid

  • MDL number MFCD23935625
  • Cas Number : 1520767-28-5
  • Molecular Weight : 385.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-657719

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-657719
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart