ENA947407042 Enamine

4-bromo-3-(bromomethyl)-1-methyl-1H-pyrazole hydrobromide

  • MDL number MFCD31381350
  • Cas Number : 2126177-73-7
  • Molecular Weight : 334.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-643001

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-643001
Availability details for
Quantity
  • Availability
  • Add to Cart