ENA947407849 Enamine

5-bromo-2-(4,5-dihydro-1,3-oxazol-2-yl)aniline

  • MDL number MFCD30751315
  • Cas Number : 1807700-37-3
  • Molecular Weight : 241.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-392839

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-392839
Availability details for
Quantity
  • Availability
  • Add to Cart