ENA947407857 Enamine

9-benzyl-1-oxa-9-azaspiro[5.5]undecan-4-amine dihydrochloride

  • MDL number MFCD19103509
  • Cas Number : 1332531-13-1
  • Molecular Weight : 333.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-392850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-392850
Availability details for
Quantity
  • Availability
  • Add to Cart