ENA947407902 Enamine

3-[5-(methoxymethyl)furan-2-yl]morpholine

  • MDL number MFCD30751334
  • Cas Number : 1596611-73-2
  • Molecular Weight : 197.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-392980

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-392980
Availability details for
Quantity
  • Availability
  • Add to Cart