ENA947407912 Enamine

N4,N6-dimethylpyrimidine-4,6-diamine

  • MDL number MFCD19218086
  • Cas Number : 66131-72-4
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-392997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-392997
Availability details for
Quantity
  • Availability
  • Add to Cart