ENA947408426 Enamine

2-(iodomethyl)-6-(trifluoromethyl)oxane

  • MDL number MFCD31381359
  • Cas Number : 2126160-29-8
  • Molecular Weight : 294.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-395684

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-395684
Availability details for
Quantity
  • Availability
  • Add to Cart