ENA947408893 Enamine

{[1-(2-methylpropyl)cyclopentyl]methyl}urea

  • MDL number MFCD23850143
  • Cas Number : 1537318-44-7
  • Molecular Weight : 198.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-396123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-396123
Availability details for
Quantity
  • Availability
  • Add to Cart