ENA947409021 Enamine

3-cyclobutyl-1,4-thiazepane

  • MDL number MFCD30751492
  • Cas Number : 1781164-65-5
  • Molecular Weight : 171.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-396479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-396479
Availability details for
Quantity
  • Availability
  • Add to Cart