ENA947409032 Enamine

2-ethylquinolin-3-ol

  • MDL number MFCD28337028
  • Cas Number : 811432-23-2
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-396501

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-396501
Availability details for
Quantity
  • Availability
  • Add to Cart