ENA947409082 Enamine

1-(chloromethyl)-2-(ethylsulfanyl)benzene

  • MDL number MFCD20623171
  • Cas Number : 23010-33-5
  • Molecular Weight : 186.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-396693

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-396693
Availability details for
Quantity
  • Availability
  • Add to Cart