ENA947409902 Enamine

4,4-difluoropentane-1-sulfonamide

  • MDL number MFCD30750329
  • Cas Number : 2126164-29-0
  • Molecular Weight : 187.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-380898

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-380898
Availability details for
Quantity
  • Availability
  • Add to Cart