ENA947410367 Enamine

methyl 2,3-dichloro-3-oxopropanoate

  • MDL number MFCD20623164
  • Cas Number : 60180-62-3
  • Molecular Weight : 170.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-396759

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-396759
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart