ENA947410630 Enamine

(1,3-benzothiazol-2-ylsulfanyl)amine

  • MDL number MFCD00267709
  • Cas Number : 2801-21-0
  • Molecular Weight : 182.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122-124
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-399413

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-399413
Availability details for
Quantity
  • Availability
  • Add to Cart