ENA947410635 Enamine

2-(3-chloro-4-methylphenyl)-2,3-dihydro-1H-isoindol-1-imine

  • MDL number MFCD02697152
  • Cas Number : 325741-03-5
  • Molecular Weight : 256.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-399421

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-399421
Availability details for
Quantity
  • Availability
  • Add to Cart