ENA947410669 Enamine

{4-[(dimethylamino)methyl]-1,3-thiazol-2-yl}methanamine

  • MDL number MFCD11934622
  • Cas Number : 933742-14-4
  • Molecular Weight : 171.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-399489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-399489
Availability details for
Quantity
  • Availability
  • Add to Cart