ENA947411212 Enamine

1-[(tert-butoxy)carbonyl]-4-methoxy-7-methyl-2,3-dihydro-1H-indole-2-carboxylic acid

  • MDL number MFCD31427917
  • Cas Number : 2137471-19-1
  • Molecular Weight : 307.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-685098

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-685098
Availability details for
Quantity
  • Availability
  • Add to Cart