ENA947411944 Enamine

4-[1-(aminomethyl)cyclobutyl]oxan-4-ol

  • MDL number MFCD21803957
  • Cas Number : 1499910-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-683228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-683228
Availability details for
Quantity
  • Availability
  • Add to Cart