ENA947415744 Enamine

(5-cyclopropyloxolan-2-yl)methanamine hydrochloride

  • MDL number MFCD30752270
  • Cas Number : 2126177-10-2
  • Molecular Weight : 177.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-398867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-398867
Availability details for
Quantity
  • Availability
  • Add to Cart