ENA947416095 Enamine

methyl 2-[(chloromethyl)sulfanyl]pyrimidine-4-carboxylate

  • MDL number MFCD30752523
  • Cas Number : 2126178-52-5
  • Molecular Weight : 218.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-399864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-399864
Availability details for
Quantity
  • Availability
  • Add to Cart