ENA947416376 Enamine

2-(3,4-difluorophenyl)-2,3-dihydro-1H-isoindol-1-imine

  • MDL number MFCD06327687
  • Cas Number : 717119-59-0
  • Molecular Weight : 244.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-399836

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-399836
Availability details for
Quantity
  • Availability
  • Add to Cart