ENA947417902 Enamine

(1R)-1-(4-fluoro-2-methylphenyl)ethan-1-ol

  • MDL number MFCD24079377
  • Cas Number : 1568151-21-2
  • Molecular Weight : 154.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-697345

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-697345
Availability details for
Quantity
  • Availability
  • Add to Cart