ENA947419395 Enamine

3-(cyclopropylamino)-4,4,4-trifluorobutanenitrile

  • MDL number MFCD31380945
  • Cas Number : 2110133-57-6
  • Molecular Weight : 178.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-701400

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-701400
Availability details for
Quantity
  • Availability
  • Add to Cart