ENA947422333 Enamine

4-(trifluoromethyl)isoquinoline

  • MDL number MFCD18418136
  • Cas Number : 120568-08-3
  • Molecular Weight : 197.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-702486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-702486
Availability details for
Quantity
  • Availability
  • Add to Cart