ENA947425017 Enamine

2-methoxy-1-azaspiro[4.5]dec-1-ene

  • MDL number MFCD31428811
  • Cas Number : 99188-43-9
  • Molecular Weight : 167.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-686904

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-686904
Availability details for
Quantity
  • Availability
  • Add to Cart