ENA947425329 Enamine

2,4-dichlorobenzoyl cyanide

  • MDL number MFCD25962596
  • Cas Number : 35022-43-6
  • Molecular Weight : 200.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-80
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-687017

Safety Information

signalword Danger
pcodes P264,P270,P301+P310,P330,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-687017
pictograms GHS06
hstatements H300
Availability details for
Quantity
  • Availability
  • Add to Cart