ENA964467358 Enamine

3-bromo-5-fluoro-2-methylbenzoic acid

  • MDL number MFCD19440362
  • Cas Number : 1187318-52-0
  • Molecular Weight : 233.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-214648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214648
Availability details for
Quantity
  • Availability
  • Add to Cart