ENA964468296 Enamine

3-{[(tert-butoxy)carbonyl]amino}-2-cycloheptylpropanoic acid

  • MDL number MFCD31414851
  • Cas Number : 2137780-07-3
  • Molecular Weight : 285.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-637490

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-637490
Availability details for
Quantity
  • Availability
  • Add to Cart