ENA964468525 Enamine

2-chloro-5-fluoropyridine-4-carbaldehyde

  • MDL number MFCD04972411
  • Cas Number : 884494-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-399760

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-399760
Availability details for
Quantity
  • Availability
  • Add to Cart