ENA964469802 Enamine

1,4-dioxa-9-azaspiro[5.6]dodecane hydrochloride

  • MDL number MFCD31433991
  • Cas Number : 2137458-11-6
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-702806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-702806
Availability details for
Quantity
  • Availability
  • Add to Cart