ENA964470386 Enamine

1-(1-methyl-1H-pyrazol-4-yl)-6-oxopiperidine-3-carboxylic acid

  • MDL number MFCD16731991
  • Cas Number : 1249816-97-4
  • Molecular Weight : 223.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-702642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-702642
Availability details for
Quantity
  • Availability
  • Add to Cart