ENA964470401 Enamine

N-methoxy-N-methylpiperidin-4-amine

  • MDL number MFCD21932718
  • Cas Number : 1517779-75-7
  • Molecular Weight : 144.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-702732

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-702732
Availability details for
Quantity
  • Availability
  • Add to Cart