ENA964470596 Enamine

4-{[(tert-butoxy)carbonyl]amino}-1-methyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD26516636
  • Cas Number : 1506512-05-5
  • Molecular Weight : 241.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-697336

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-697336
Availability details for
Quantity
  • Availability
  • Add to Cart