ENA964471484 Enamine

6-fluoro-2-propoxyquinoline-3-carboxylic acid

  • MDL number MFCD31434540
  • Cas Number : 2137600-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-703747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-703747
Availability details for
Quantity
  • Availability
  • Add to Cart