ENA964471825 Enamine

tert-butyl 4-(2-methoxy-2-oxoethyl)-4-(oxiran-2-yl)piperidine-1-carboxylate

  • MDL number MFCD31434920
  • Cas Number : 2138158-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-704516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-704516
Availability details for
Quantity
  • Availability
  • Add to Cart