ENA964472340 Enamine

2-(thian-4-yloxy)ethan-1-ol

  • MDL number MFCD28133024
  • Cas Number : 113194-34-6
  • Molecular Weight : 162.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-704158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-704158
Availability details for
Quantity
  • Availability
  • Add to Cart