ENA964473547 Enamine

2-cyclopropyl-4-fluorobenzoic acid

  • MDL number MFCD16778838
  • Cas Number : 959992-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-705532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-705532
Availability details for
Quantity
  • Availability
  • Add to Cart