ENA964473587 Enamine

(1R,2R)-2-ethylcyclopropane-1-carboxylic acid

  • MDL number MFCD24502412
  • Cas Number : 1923743-63-8
  • Molecular Weight : 114.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-705382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-705382
Availability details for
Quantity
  • Availability
  • Add to Cart