ENA964473603 Enamine

(1R,2R)-2-(methoxymethyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD31435705
  • Cas Number : 2137069-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-705386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-705386
Availability details for
Quantity
  • Availability
  • Add to Cart