ENA964477505 Enamine

4-(trimethylsilyl)benzoic acid

  • MDL number MFCD00092492
  • Cas Number : 15290-29-6
  • Molecular Weight : 194.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-714511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-714511
Availability details for
Quantity
  • Availability
  • Add to Cart