ENA964480258 Enamine

(4,5,6,7-tetrahydro-1H-1,3-benzodiazol-2-yl)methanamine; trifluoroacetic acid

  • MDL number MFCD31438838
  • Cas Number : 2138135-70-1
  • Molecular Weight : 265.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-716331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-716331
Availability details for
Quantity
  • Availability
  • Add to Cart