ENA964480697 Enamine

4-bromo-1-(pyrimidin-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD23881380
  • Cas Number : 1542113-05-2
  • Molecular Weight : 240.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-716136

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-716136
Availability details for
Quantity
  • Availability
  • Add to Cart