ENA964480732 Enamine

ethyl 2-(5-bromothiophen-2-yl)-2,2-difluoroacetate

  • MDL number MFCD30529730
  • Cas Number : 1286792-85-5
  • Molecular Weight : 285.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-716245

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-716245
Availability details for
Quantity
  • Availability
  • Add to Cart